missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxytetracycline Glucose Yeast Extract Agar

Product Code. 12922598
Change view
Click to view available options
Description:
Oxytetracycline Glucose Yeast Extract Agar
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description unitSize
12922598 Oxytetracycline Glucose Yeast Extract Agar Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12922598 Supplier Thermo Scientific™ Supplier No. PO0183A

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Selectively enumerate yeasts and molds from foodstuffs with this selective, ready-to-use medium.

Selectively enumerate yeasts and molds from foodstuffs with Thermo Scientific™ Oxoid™ Oxytetracycline Glucose Yeast Extract Agar1,2.The medium is based upon the formulation described by Mossel et al.3 which utilizes oxytetracycline selectivity to provide increased counts of yeasts and molds compared to other media that utilize a low pH as a selective agent. Improve the recovery of physically stressed yeast cells on this neutral pH medium4.

Use Oxytetracycline Glucose Yeast Extract Agar for selection and enumeration of yeasts and molds from foodstuffs.

  • Increased colony count of yeasts and molds compared to that of low pH media.
  • Improved recovery of physically stressed yeast cells.
  • Ready-to-use convenience of prepared media.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Color Straw
Content And Storage 2°C to 10°C
Description Oxytetracycline Glucose Yeast Extract Agar
Format 90mm Monoplate
Manufacturing Origin United Kingdom
Product Type Agar
Fill Weight 19g±2.0.g
Packaging Type 10 x 90mm plates, film wrapped
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.