Learn More
Thermo Scientific™ Oxoid™ VCNT Selective Supplement
The Oxoid VCNT Selective Supplement is used for the preparation of Thayer Martin Medium for the isolation of pathogenic Neisseria spp.
Brand: Thermo Scientific™ SR0091E
Code : 65
Additional Details : Weight : 0.15510kg
Description
Oxoid VCNT Selective Supplement is used for the preparation of Thayer Martin Medium for the isolation of pathogenic Neisseria spp.
- Add to GC Agar Base, Part No. CM0367B
Specifications
VCNT Selective Supplement | |
Selective Supplement | |
Each vial for 500mL medium |
Freeze-dried | |
10/Pk. |