missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Oxoid™ VCNT Selective Supplement
The Oxoid VCNT Selective Supplement is used for the preparation of Thayer Martin Medium for the isolation of pathogenic Neisseria spp.
£139.00 - £291.00
Specifications
Description | VCNT Selective Supplement |
---|---|
Form | Freeze-dried |
Product Type | Selective Supplement |
Quantity | 10/Pk. |
Product Code | Brand | Yield | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Yield | Price | Quantity & Availability | |||||
11434169
|
Thermo Scientific™
SR0091E |
Each vial for 500mL medium |
£139.00
Pack of 10 |
In Stock
Log in to see stock. |
|||||
11444169
|
Thermo Scientific™
SR0091H |
Each vial for 2L medium |
£291.00
Pack of 10 |
In Stock
Log in to see stock. |
|||||
Description
Oxoid VCNT Selective Supplement is used for the preparation of Thayer Martin Medium for the isolation of pathogenic Neisseria spp.
- Add to GC Agar Base, Part No. CM0367B
Specifications
VCNT Selective Supplement | |
Freeze-dried | |
Selective Supplement | |
10/Pk. |