missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Prepared Yeast Extract Agar Bottled Media, 498-502 g, 500 mL sirop bottle, 10 bottles

Product Code. 15321292
Change view
Click to view available options
Fill Weight:
100.0 to 101.5 g
20 to 20.3 g
250 to 254 g
498 to 502 g
Format:
125 mL sirop bottle
300 mL Sirop Bottle
500 mL Sirop Bottle
Universal Bottle
Unit Size:
1000mL
500mL
Pack of 10
Pack of 24
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Fill Weight Format unitSize
15321292 250 to 254 g 300 mL Sirop Bottle Pack of 10
15264867 100.0 to 101.5 g 125 mL sirop bottle 1000mL
11923782 100.0 to 101.5 g 125 mL sirop bottle 1000mL
12756989 20 to 20.3 g Universal Bottle Pack of 24
11932622 498 to 502 g 500 mL Sirop Bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 15321292 Supplier Thermo Scientific™ Supplier No. BO0556T

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Enumerate microorganisms in potable and freshwater samples with this agar.

Yeast Extract Agar is used for the enumeration of microorganisms in potable and freshwater samples.

TRUSTED_SUSTAINABILITY

Specifications

Color Straw
Content And Storage 2°C to 25°C
Format 300 mL Sirop Bottle
Manufacturing Origin United Kingdom
Product Type Agar
Fill Weight 250 to 254 g
Packaging Type 10 Sirop Bottles with Screw Cap Closures
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.