missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid MRS Agar

Product Code. 10108502
Click to view available options
Color:
Ivory, transparent
Straw, clear
Manufacturing Origin:
Germany
United Kingdom
Fill Weight:
17g ± 5%
19.5g ± 1.0g
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 10108502

Brand: Thermo Scientific™ PO5047A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cultivate, isolate and enumerate lactic acid bacteria with Thermo Scientific™ Oxoid™ MRS Agar.

Cultivate, isolate and enumerate lactic acid bacteria with Thermo Scientific™ Oxoid™ MRS Agar. The formulation was developed by de Man, Rogosa and Sharpe1 to replace a variable product (tomato juice) and, at the same time, to provide a medium which would support good growth of lactobacilli in general, even those strains which showed poor growth in existing media. It gives more profuse growth of all strains of lactobacilli, especially the difficult and slow-growing strains of Lactobacillus brevis and Lactobacillus fermenti1,2.

MRS medium is superior to the tomato juice medium of Briggs2 and the meat extract tomato juice medium of de Man. MRS Agar was designed to encourage the growth of the lactic acid bacteria which includes species of the following genera: Lactobacillus, Streptococcus, Pediococcus and Leuconostoc. All these species can produce lactic acid in considerable amounts. They are Gram +ve, catalase and oxidase –ve and are fastidious in their nutritional requirements. Growth is enhanced considerably by micro-aerobic conditions.

Generally the lactic acid bacteria show delayed growth and smaller colony size than other microorganisms. They may be overgrown in non-selective media, especially if incubation is required for 2 to 4 days.

Selection can be made by pH adjustment; thus, lactobacilli will tolerate lower pH levels than streptococci (pH 5.0-6.5), with pediococci and leuconostocs growing best within this range. Inhibitors of the main groups of competitor microflora include thallous acetate, sodium acetate, sorbic acid, acetic acid, sodium nitrite, cycloheximide and polymyxin. These substances can be used at varying concentrations and combinations, but inevitably a compromise has to be reached between selectivity and productivity of the organism sought3.

MRS Agar with sorbic acid has been described3,4. This is MRS medium with its pH reduced to 5.7 and 0.14% w/v sorbic acid added (=0.2% w/v potassium sorbate).

An evaluation of media for selective enumeration of Lactobacillus acidophilus and Bifidobacterium species showed that minor adjustments to the basic formula of MRS Agar can readily be made to optimize its

performance for determining the content of Lactobacillus acidophilus and Bifidobacterium spp. in the presence of other lactic acid bacteria which are in yoghurt.

The lactobacilli are micro-aerophilic and generally require layer plates for aerobic cultivation on solid media.

Submerged or surface colonies may be compact or feathery, and are small, opaque and white3,4.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

TRUSTED_SUSTAINABILITY

Specifications

Color Ivory, transparent
Content And Storage 6°C to 12°C
Description Oxoid M.R.S. Agar
Format 90 mm Monoplate
Manufacturing Origin Germany
Fill Volume 17 mL
Fill Weight 17g ± 5%
Packaging Type Flow Wrapped - Single Layer
Quantity 10/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.