missing translation for 'onlineSavingsMsg'
Learn More

Thermo Fisher Scientific™ Oxoid™ Eosine Methylene Blue Agar (modified) Levine

Product Code. 10148452
Change view
Click to view available options
Fill Weight:
17 g ±5%
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Fill Weight Unit Size
10148452 17 g ±5% Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10148452 Supplier Thermo Fisher Scientific™ Supplier No. PO5045A
  • ONLINE EXCLUSIVE PRICE
    £21.45
    £20.14 / Pack of 10
    Save £1.31
    6% Off
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

An isolation medium for the differentiation of Enterobacteriaceae

TRUSTED_SUSTAINABILITY

Specifications

Color Pearl ruby red, transparent
Content And Storage 6 to 12°C, dark
Form Agar
Format Poured plate
Media Type Eosine Methylene Blue Agar (modified) Levine
Product Type Microbiology Media
Fill Weight 17 g ±5%
For Use With (Application) An isolation medium for the differentiation of Enterobacteriaceae
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.