missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Dip-Slides Clinical

Product Code. 10055732
Change view
Click to view available options
Quantity:
10/Pk.
Format:
Dip Slide
Media Type:
MacConkey Agar and CLED Medium
Manufacturing Origin:
Italy
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Format Media Type Manufacturing Origin unitSize
10055732 10/Pk. Dip Slide MacConkey Agar and CLED Medium Italy Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10055732 Supplier Thermo Scientific™ Supplier No. GFD01A
Reliable Agri Testing Solutions
Use promo code 28024
View all promotions
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Make colony counting for the bacterial assessment of urine simple with the easily disposable Thermo Scientific™ Oxoid™ Dip-Slides Clinical.

Make colony counting for the bacterial assessment of urine simple with the easily disposable Thermo Scientific™ Oxoid™ Dip-Slides™ Clinical. Simple and safe for an unskilled operator to use, these dip slides are coated on both sides with a solid culture medium with the opportunity for different media to be carried on each side, making the process time- and cost-effective. This sterile dip slide can be inoculated in the ward or doctor's surgery and then returned to its plastic container for easy transport back to the laboratory.

Previous methods of detecting urinary tract infections depended on freshly voided urine being delivered promptly to a bacteriological laboratory for examination, either immediately or after a short period of refrigerated storage. The dip slide was developed as a logical extension of this method, using a microscope slide coated on each side with a different culture medium. With these dip slides, larger areas of media can be used for bacterial assessment of urine, making them time- and cost-effective.

  • Slide is made of easily disposable plastic with raised edges to ensure an even thickness of culture medium
  • Molded grid allows for easier colony counting
  • Slide is shaped to ensure drainage of fluid from the slide
  • Cap forms a convenient handle by which it may be held without risk of touching the culture medium
  • Bacteria chart is included, make examining growth simple for the operator and where laboratory facilities are limited

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products with a comprehensive line of media and diagnostic products, providing a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 15°C to 25°C
Format Dip Slide
Manufacturing Origin Italy
Media Type MacConkey Agar and CLED Medium
Product Type Dip Slide
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.