missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Dichloran Rose Bengal Chloramphenicol

Product Code. 15581984
Click to view available options
Quantity:
10 Pcs/Pk.
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15581984

Brand: Thermo Scientific™ PO1278A

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Achieve precise enumeration of yeasts and molds with this agar, ideal for food and environmental samples.

Achieve precise enumeration of yeasts and molds with Dichloran Rose Bengal Chloramphenicol Agar, ideal for food and environmental samples.

TRUSTED_SUSTAINABILITY

Specifications

Color Straw
Content And Storage 2°C to 12°C
Format 90 mm Plate
Manufacturing Origin United Kingdom
Product Type Agar Plate
Fill Weight 17 g
Quantity 10 Pcs/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.