Learn More
Thermo Scientific™ Contrast™ MRSA Broth, 3 ± 0,3mL
Description
Improve laboratory productivity by adding multiple samples from one patient to one tube and achieve results within 18 to 24 hours with Thermo Scientific™ Contrast™ MRSA Broth. Rapidly eliminate negative results and confirm positive results using Thermo Scientific™ Brilliance™ MRSA 2 Agar.
Simplicity and speed are important factors when screening for MRSA, as reliable results are necessary for an effective infection control process.
Contrast MRSA Broth contains the same antibiotic marker used in Brilliance MRSA Agar to select for MRSA strains. Other antibiotics and inhibitory agents are present to inhibit non-MRSA and Gram-negative organisms. The broth contains an indicator dye, phenol red, and two sugars (mannitol and trehalose) which are fermented by MRSA strains. This causes a decrease in the pH of the broth, resulting in a color change from red to orange/yellow, indicating a presumptive positive result. The improved broth contains a low level of nalidixic acid, which has activity against both Gram-positive and Gram-negative bacteria. The concentration is optimized for maximum activity against non-target organisms without compromising growth of methicillin-resistant S. aureus, including ciprofloxicin-sensitive strains. Contrast MRSA Broth can be inoculated from screening swabs taken from hospital patients or staff, according to local guidelines. Multiple swabs (maximum four) from the same patient can be pooled into a single broth.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Specifications
| Color | Red, clear |
| Content And Storage | 2°C to 8°C |
| Description | Contrast MRSA Broth |
| Format | Universal |
| Manufacturing Origin | United Kingdom |
| Product Type | Microbiology Media |
| Fill Volume | 3 mL |
| Fill Weight | 3 ± 0,3mL |
| Packaging Type | 24 universal in a carton box |
| Quantity | 24/Pk. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.