missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Brain Heart Infusion Broth

Product Code. 12756959
Click to view available options
Fill Weight:
100 to 101.5 g
9.0-9.3g
Format:
125 mL Sirop Bottle
Universal bottle
Unit Size:
Pack of 10
Pack of 24
This item is not returnable. View return policy

Product Code. 12756959

Brand: Thermo Scientific™ BO1230D

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A highly nutritious, general-purpose medium for the growth of fastidious microorganisms from clinical samples.

Brain Heart Infusion Broth is a highly nutritious, general-purpose medium for the growth of fastidious microorganisms from clinical samples.

TRUSTED_SUSTAINABILITY

Specifications

Color Straw
Content And Storage 2 to 25°C
Format Universal bottle
Manufacturing Origin United Kingdom
Product Type Broth
Fill Weight 9.0-9.3g
Packaging Type Twenty-four universal bottles with screw cap closures in a box.
Quantity 24/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.