missing translation for 'onlineSavingsMsg'
Learn More

Oxcarbazepine, 98%

Product Code. 15476478
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15476478 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15476478 Supplier Thermo Scientific Alfa Aesar Supplier No. H60014.MD
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 28721-07-5 | C15H12N2O2 | 252.273 g/mol

Oxcarbazepine is a sodium channel protein inhibitor. It is an anticonvulsant and mood-stab.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in DMSO, methanol, water, ethanol and acetone.

Notes
Store at -20°C.
TRUSTED_SUSTAINABILITY

Specifications

CAS 28721-07-5
Molecular Formula C15H12N2O2
MDL Number MFCD00865307
Quantity 250 mg
Synonym oxcarbazepine, trileptal, oxcarbamazepine, oxacarbazepine, oxcarbazepina, oxcarbazepinum, timox, oxcarbazepinum inn-latin, oxcarbazepina inn-spanish, oxcarbazepine inn
Solubility Information Soluble in DMSO,methanol,water,ethanol and acetone.
InChI Key CTRLABGOLIVAIY-UHFFFAOYSA-N
SMILES C1C2=CC=CC=C2N(C3=CC=CC=C3C1=O)C(=O)N
IUPAC Name 5-oxo-6H-benzo[b][1]benzazepine-11-carboxamide
Molecular Weight (g/mol) 252.273
PubChem CID 34312
ChEBI CHEBI:7824
Formula Weight 252.27
Percent Purity 98%
Chemical Name or Material Oxcarbazepine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.