missing translation for 'onlineSavingsMsg'
Learn More

Oxamide, 98+%

Product Code. 11404633
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11404633 500 g 500g
11494623 100 g 100g
11414633 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11404633 Supplier Thermo Scientific Alfa Aesar Supplier No. A11570.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 471-46-5 | C2H4N2O2 | 88.066 g/mol

Oxamide is used as a stabilizer for nitrocellulose preparations. It also finds use in APCP rocket motors as a high performance burn rate suppressant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Oxamide is used as a stabilizer for nitrocellulose preparations. It also finds use in APCP rocket motors as a high performance burn rate suppressant.

Solubility
Soluble in ethanol, slightly soluble in water and insoluble in diethyl ether.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 471-46-5
Molecular Formula C2H4N2O2
Molecular Weight (g/mol) 88.066
MDL Number MFCD00008007
InChI Key YIKSCQDJHCMVMK-UHFFFAOYSA-N
Synonym ethanediamide, oxalamide, oxamimidic acid, oxalic acid diamide, diaminoglyoxal, oxamid, amid kyseliny stavelove, formimidic acid, 1-carbamoyl, oxamid czech, unii-sbe4m0223e
PubChem CID 10113
ChEBI CHEBI:48248
IUPAC Name oxamide
SMILES C(=O)(C(=O)N)N

Specifications

Melting Point ∼350°C (decomposition)
Density 1.667
Odor Odorless
Quantity 500 g
Beilstein 1743262
Merck Index 14,6918
Solubility Information Soluble in ethanol,slightly soluble in water and insoluble in diethyl ether.
Formula Weight 88.07
Percent Purity ≥98%
Chemical Name or Material Oxamide
compliance-icons
Product Identifier
  • Oxamide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Oxamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.