accessibility menu, dialog, popup

UserName

CAS RN 471-46-5

H2NONH2O

CAS RN 471-46-5

IUPAC Name: ethanediamide
Synonyms: oxamide 1-carbamoylformimidic acid amid kyseliny stavelove oxamimidic acid
Molecular Weight (g/mol): 88.07
Molecular Formula: C2H4N2O2
InChi Key: YIKSCQDJHCMVMK-UHFFFAOYSA-N
SMILES: NC(=O)C(N)=O
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)