missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OSGEPL1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£149.00 - £366.00
Specifications
| Antigen | OSGEPL1 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50 - 1:100, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18679640
|
Novus Biologicals
NBP2-93751-0.02ml |
0.02 mL |
£149.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18625561
|
Novus Biologicals
NBP2-93751-0.1ml |
0.1 mL |
£366.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
OSGEPL1 Polyclonal antibody specifically detects OSGEPL1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| OSGEPL1 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| EC 3.4.24.57, O-sialoglycoprotein endopeptidase like 1, O-sialoglycoprotein endopeptidase-like 1, O-sialoglycoprotein endopeptidase-like protein 1, probable O-sialoglycoprotein endopeptidase 2, putative sialoglycoprotease type 2, t(6)A37 threonylcarbamoyladenosine biosynthesis protein OSGEPL1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-270 of human OSGEPL1 (NP_071748.2). MLILTKTAGVFFKPSKRKVYEFLRSFNFHPGTLFLHKIVLGIETSCDDTAAAVVDETGNVLGEAIHSQTEVHLKTGGIVPPAAQQLHRENIQRIVQEALSASGVSPSDLSAIATTIKPGLALSLGVGLSFSLQLVGQLKKPFIPIHHMEAHALTIRLTNKVEFPFLVLLISGGHCLLALVQGVSDFLLLGKSLDIAPGDMLDKVARRLSLIKHPECSTMSGGKAIEHLAKQGNRFHFDIKPPLHHAKNCDFSFTGLQHVTDKIIMKKEKE | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50 - 1:100, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 64172 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title