missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OSGEPL1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93751-0.1ml
This item is not returnable.
View return policy
Description
OSGEPL1 Polyclonal antibody specifically detects OSGEPL1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| OSGEPL1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50 - 1:100, Immunohistochemistry-Paraffin | |
| EC 3.4.24.57, O-sialoglycoprotein endopeptidase like 1, O-sialoglycoprotein endopeptidase-like 1, O-sialoglycoprotein endopeptidase-like protein 1, probable O-sialoglycoprotein endopeptidase 2, putative sialoglycoprotease type 2, t(6)A37 threonylcarbamoyladenosine biosynthesis protein OSGEPL1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-270 of human OSGEPL1 (NP_071748.2). MLILTKTAGVFFKPSKRKVYEFLRSFNFHPGTLFLHKIVLGIETSCDDTAAAVVDETGNVLGEAIHSQTEVHLKTGGIVPPAAQQLHRENIQRIVQEALSASGVSPSDLSAIATTIKPGLALSLGVGLSFSLQLVGQLKKPFIPIHHMEAHALTIRLTNKVEFPFLVLLISGGHCLLALVQGVSDFLLLGKSLDIAPGDMLDKVARRLSLIKHPECSTMSGGKAIEHLAKQGNRFHFDIKPPLHHAKNCDFSFTGLQHVTDKIIMKKEKE | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 64172 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction