missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OSBPL5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-38858
This item is not returnable.
View return policy
Description
OSBPL5 Polyclonal specifically detects OSBPL5 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| OSBPL5 | |
| Polyclonal | |
| Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Q9H0X9 | |
| OSBPL5 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: LCGLPASATVHPDQDLFPLNGSSLENDAFSDKSERENPEESDTETQDHSRKTESGSDQSETPGAPVRRGTTYVEQVQEEL | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| EC 1.2.1, EC 2.7.7.6, FLJ42929, KIAA1534oxysterol-binding protein-related protein 5, OBPH1, ORP-5, ORP5oxysterol-binding protein homologue 1, OSBP-related protein 5, oxysterol binding protein-like 5, oxysterol-binding protein homolog 1, Oxysterol-binding protein homolog 1; | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 114879 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction