missing translation for 'onlineSavingsMsg'
Learn More

Orotic acid monohydrate, 98%

Product Code. 10160122 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10160122 5 g Glass Bottle 5g
10307160 100 g Plastic bottle 100g
10539243 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10160122 Supplier Thermo Scientific Acros Supplier No. 129570050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 50887-69-9 | C5H4N2O4 | 156.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50887-69-9
Molecular Formula C5H4N2O4
Molecular Weight (g/mol) 156.10
MDL Number MFCD00006027
InChI Key PXQPEWDEAKTCGB-UHFFFAOYSA-N
Synonym orotic acid monohydrate, orotic acid hydrate, 2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylic acid hydrate, lactinium, oroticum acidum, 2,6-dioxo-1,2,3,6-tetrahydro-4-pyrimidinecarboxylic acid, orotc acd hydrate?, uracil-6-carboxylic acid monohydrate, uracil-4-carboxylic acid hydrate
PubChem CID 16218504
IUPAC Name 2,4-dioxo-1H-pyrimidine-6-carboxylic acid;hydrate
SMILES OC(=O)C1=CC(=O)NC(=O)N1

Specifications

Melting Point 334°C
Color White
Loss on Drying 12.0% max. (1 g, 130°C, 3 hrs)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Quantity 5 g
Beilstein 25, 253
Merck Index 15, 6972
Solubility Information Solubility in water: 1.3g/L (20 c). Other solubilities: insoluble in chloroform
Formula Weight 174.11
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Orotic acid monohydrate
Show More Show Less
compliance-icons
Product Identifier
  • Orotic acid monohydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.