accessibility menu, dialog, popup

UserName

HOOONHOHN

CAS RN 50887-69-9

IUPAC Name: 2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylate
Synonyms: orotate
Molecular Weight (g/mol): 155.09
Molecular Formula: C5H3N2O4
InChi Key: PXQPEWDEAKTCGB-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC(=O)NC(=O)N1
Molecular Weight (g/mol) 
  • (5)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (5)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)