missing translation for 'onlineSavingsMsg'
Learn More

Orotic acid hydrate, 98%

Product Code. 11438517
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11438517 50 g 50g
11448517 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11438517 Supplier Thermo Scientific Alfa Aesar Supplier No. A18594.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 50887-69-9 | C5H4N2O4 | 156.10 g/mol

Orotic acid hydrate is used in cosmetic and food industry. It is also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Orotic acid hydrate is used in cosmetic and food industry. It is also used as a pharmaceutical intermediate.

Solubility
Soluble in water (1.7 mg/ml), and ammonium hydroxide. Insoluble in chloroform.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50887-69-9
Molecular Formula C5H4N2O4
Molecular Weight (g/mol) 156.10
MDL Number MFCD00006027
InChI Key PXQPEWDEAKTCGB-UHFFFAOYSA-N
Synonym orotic acid monohydrate, orotic acid hydrate, 2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylic acid hydrate, lactinium, oroticum acidum, 2,6-dioxo-1,2,3,6-tetrahydro-4-pyrimidinecarboxylic acid, orotc acd hydrate?, uracil-6-carboxylic acid monohydrate, uracil-4-carboxylic acid hydrate
PubChem CID 16218504
IUPAC Name 2,4-dioxo-1H-pyrimidine-6-carboxylic acid;hydrate
SMILES OC(=O)C1=CC(=O)NC(=O)N1

Specifications

Melting Point >300°C
Quantity 50 g
Beilstein 383901
Merck Index 14,6876
Solubility Information Soluble in water (1.7mg/ml),and ammonium hydroxide. Insoluble in chloroform.
Formula Weight 156.10 (Anhydrous)
Percent Purity 98%
Chemical Name or Material Orotic acid hydrate
compliance-icons
Product Identifier
  • Orotic acid hydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Orotic acid hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.