missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ AQ3700 AQUAfast™ Colorimeter

Product Code. p-4532054
Click to view available options
Type:
Colorimeter
Sealing Ring for 24 mm Vial
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15761617

Brand: Thermo Scientific™ AC3SR24

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Select this multiparameter colorimeter to manage your water and wastewater analysis using over 90 preprogrammed methods.

Select the Thermo Scientific™ Orion™ AQUAfast AQ3700 Colorimeter to manage your water and wastewater analysis needs. This instrument has over 90 pre-programmed methods for common test parameters with automatic selection of 6 LED wavelengths and countdown timer. Methods include total nitrogen and total phosphate nutrient testing or create up to 10 custom methods using 2-14 standards and blank. This IP67 waterproof meter records 1,000 data points, offers a long battery life of about 26 hours continuous use or 3,500 tests, and multi-language user interface in English, French, German, Italian, Polish, Portuguese and Spanish.

Features:

  • Wide range of disinfectants in various formats to broaden our offering to a variety of industries
  • Large graphic display with step-by-step user instructions
  • Automatic countdown to measurement
  • Consistent results from shift to shift
  • Selectable languages of English, French, German, Italian, Spanish, Portuguese (Brazilian), and Polish with other languages uploadable via RJ 45 connection

Requires:

  • 4 AA batteries

Recommended for:

  • Wastewater
  • Drinking water
  • Nutrient monitoring
  • Environmental testing
  • Food and beverage
  • Industrial applications
Note: For the most accurate results, use only Thermo Scientific Orion reagents.
TRUSTED_SUSTAINABILITY

Specifications

Type Sealing Ring for 24 mm Vial
For Use With (Equipment) AQ3700 Colorimeter
Storage Requirements 5°C to 40°C

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.