missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ AQ3700 AQUAfast™ Colorimeter

Product Code. 15761617
Change view
Click to view available options
Type:
Colorimeter
Sealing Ring for 24 mm Vial
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
15761617 Sealing Ring for 24 mm Vial Each
12708725 Colorimeter Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15761617 Supplier Thermo Scientific™ Supplier No. AC3SR24
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Select this multiparameter colorimeter to manage your water and wastewater analysis using over 90 preprogrammed methods.

Select the Thermo Scientific™ Orion™ AQUAfast AQ3700 Colorimeter to manage your water and wastewater analysis needs. This instrument has over 90 pre-programmed methods for common test parameters with automatic selection of 6 LED wavelengths and countdown timer. Methods include total nitrogen and total phosphate nutrient testing or create up to 10 custom methods using 2-14 standards and blank. This IP67 waterproof meter records 1,000 data points, offers a long battery life of about 26 hours continuous use or 3,500 tests, and multi-language user interface in English, French, German, Italian, Polish, Portuguese and Spanish.

Features:

  • Wide range of disinfectants in various formats to broaden our offering to a variety of industries
  • Large graphic display with step-by-step user instructions
  • Automatic countdown to measurement
  • Consistent results from shift to shift
  • Selectable languages of English, French, German, Italian, Spanish, Portuguese (Brazilian), and Polish with other languages uploadable via RJ 45 connection

Requires:

  • 4 AA batteries

Recommended for:

  • Wastewater
  • Drinking water
  • Nutrient monitoring
  • Environmental testing
  • Food and beverage
  • Industrial applications
Note: For the most accurate results, use only Thermo Scientific Orion reagents.
TRUSTED_SUSTAINABILITY

Spezifikation

Type Sealing Ring for 24 mm Vial
For Use With (Equipment) AQ3700 Colorimeter
Storage Requirements 5°C to 40°C

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.