missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ 2117LL Low-Level Chloride Analyzer

Product Code. p-6043103
Click to view available options
Type:
Chloride Analyzer
Voltage:
230 V
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15383001

Brand: Thermo Scientific™ 2117LL230

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Get continuous online monitoring and control of steam cycle and boiler chemistry with the Thermo Scientific™ Orion™ 2117LL Low-Level Chloride Analyzer.

Get continuous online monitoring and control of steam cycle and boiler chemistry with the Thermo Scientific™ Orion™ 2117LL Low-Level Chloride Analyzer, for use with power generation applications requiring chloride measurement at trace levels.

Without accurate and real-time monitoring of chloride, the risk of corrosion fatigue, stress corrosion cracking and pitting can create significant costs in maintenance and downtime. Early detection of chloride ingress from feedwater helps your plant maintain the optimal balance of water purification with ease and peace of mind.

Features:

  • Detects chloride in real-time, providing accurate and reliable results
  • Provides high performance results with confidence
  • Includes unique electrode technology for stability and a virtually maintenance-free fluidic design

Benefits:

  • Calibration record is maintained for up to 60 days between reagent changes.
  • System is one of the only real-time monitors on the market capable of reading down to 5ppb.
  • Long-life, solid-state sensor technology offers stability and high performance.
  • No moving parts means simple operation and minimal maintenance.
  • Measurements are repeatable and verifiable.

Ordering information:

The chloride analyzer is available in several configurations including the analyzer instrument only or with a protective enclosure.

TRUSTED_SUSTAINABILITY

Specifications

Description Low Level Chloride Analyzer, 230V
Type Chloride Analyzer
Voltage 230 V
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.