missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ ORF73/HHV8 Antibody, Novus Biologicals™
Sheep Polyclonal Antibody
£162.00 - £386.00
Specifications
| Antigen | ORF73/HHV8 |
|---|---|
| Dilution | Western Blot : 1-5 μg/mL, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation : 10-20 μg/mL, Immunohistochemistry-Frozen |
| Applications | ELISA, Immunoprecipitation, Western Blot, Immunohistochemistry (Frozen), Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30067122
|
Novus Biologicals™
NBP3-30981-50ug |
50 μg |
£162.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30067003
|
Novus Biologicals™
NBP3-30981-250ug |
250 μg |
£386.00
250µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ORF73/HHV8 Polyclonal antibody specifically detects ORF73/HHV8 in Human samples. It is validated for ELISA,Immunoprecipitation,Western Blot,Immunohistochemistry (Frozen),Immunocytochemistry/ ImmunofluorescenceSpecifications
| ORF73/HHV8 | |
| ELISA, Immunoprecipitation, Western Blot, Immunohistochemistry (Frozen), Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Sheep | |
| Adaptive Immunity, Immunology, Virology Bacteria and Parasites | |
| Phosphate buffered saline | |
| 935114 | |
| IgG | |
| Ammonium sulfate precipitation |
| Western Blot : 1-5 μg/mL, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation : 10-20 μg/mL, Immunohistochemistry-Frozen | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| HHV 8, Human Herpes Virus 8, Kaposi's sarcoma associated herpes virus, KSHV | |
| Inclusion Bodies from E. Coli expressing his-tagged full length human ORF73/HHV8 protein. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title