missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ ORF73/HHV8 Antibody, Novus Biologicals™
Sheep Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30981-250ug
This item is not returnable.
View return policy
Description
ORF73/HHV8 Polyclonal antibody specifically detects ORF73/HHV8 in Human samples. It is validated for ELISA,Immunoprecipitation,Western Blot,Immunohistochemistry (Frozen),Immunocytochemistry/ Immunofluorescence
Specifications
| ORF73/HHV8 | |
| Polyclonal | |
| Western Blot : 1-5 μg/mL, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation : 10-20 μg/mL, Immunohistochemistry-Frozen | |
| HHV 8, Human Herpes Virus 8, Kaposi's sarcoma associated herpes virus, KSHV | |
| Inclusion Bodies from E. Coli expressing his-tagged full length human ORF73/HHV8 protein. | |
| 250 μg | |
| Adaptive Immunity, Immunology, Virology Bacteria and Parasites | |
| 935114 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
| ELISA, Immunoprecipitation, Western Blot, Immunohistochemistry (Frozen), Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Phosphate buffered saline | |
| Sheep | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction