missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Oregon Green™ 514 Carboxylic Acid, Succinimidyl Ester
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Description
The amine-reactive Oregon Green™ 514 succinimidyl ester can be used to create green fluorescent bioconjugates with excitation/emission maxima ∼489/526 nm.
This fluorinated analog of fluorescein overcomes some of the key limitations of fluorescein, including greater photostability and a lower pKa, making its fluorescence essentially pH insensitive in the physiological pH range.
This fluorinated analog of fluorescein overcomes some of the key limitations of fluorescein, including greater photostability and a lower pKa, making its fluorescence essentially pH insensitive in the physiological pH range.
Specifications
Specifications
| Quantity | 5 mg |
| Product Type | Succinimidyl Ester |
| Content And Storage | Store in freezer (-5 to -30°C) and protect from light. |
| Chemical Reactivity | Amine |
| Shipping Condition | Room Temperature |
| Reactive Moiety | Active Ester, Succinimidyl Ester |
| Label or Dye | Oregon Green™ 514 |
| Label Type | Classic Dyes |
| Product Line | Oregon Green |
| Emission | 526 |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction