missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Oregon Green™ 488 Azide (Oregon Green™ 6-Carboxamido-(6-Azidohexanyl), Triethylammonium Salt), 6-isomer

Product Code. 10688375
Click to view available options
Quantity:
0.5 mg
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10688375

Brand: Molecular Probes™ O10180

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Oregon Green™ 488 Azide (Oregon Green™ 6-Carboxamido-(6-Azidohexanyl), Triethylammonium Salt), 6-isomer

The green-fluorescent Oregon Greenr™ 488 azide is reactive with terminal alkynes via a copper-catalyzed click reaction. In addition to being a bright and photostable alternative to fluorescein for use in flow cytometry, microscopy and HCS, Oregon Green™ 488 can also be utilized as a bio-orthogonal or biologically unique hapten for use in applications requiring signal amplification.
TRUSTED_SUSTAINABILITY

Specifications

Format Solid
Quantity 0.5 mg
Product Type Azide
Solubility DMF (Dimethylformamide), DMSO (Dimethylsulfoxide)
Content And Storage Store at ≤-20°C, desiccate and protect from light.
Chemical Reactivity Alkyne
Detection Method Fluorescence
Shipping Condition Room Temperature
System Type Click-iT™
Reactive Moiety Amine, Azide
Label or Dye Oregon Green™ 488
Label Type Classic Dyes
Color Green
Product Line Molecular Probes, Oregon Green
Emission 524
Excitation 496
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.