missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OR2W1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-69041
This item is not returnable.
View return policy
Description
OR2W1 Polyclonal specifically detects OR2W1 in Human samples. It is validated for Western Blot.
Specifications
| OR2W1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| hs6M1-15, MGC119162, MGC119163, MGC119165, olfactory receptor 2W1, Olfactory receptor OR6-13, olfactory receptor, family 2, subfamily W, member 1 | |
| Rabbit | |
| 36 kDa | |
| 100 μL | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q9Y3N9 | |
| OR2W1 | |
| Synthetic peptides corresponding to OR2W1 (olfactory receptor, family 2, subfamily W, member 1) The peptide sequence was selected from the C terminal of OR2W1. Peptide sequence VVSMFYGTIIYMYLQPGNRASKDQGKFLTLFYTVITPSLNPLIYTLRNKD. | |
| Affinity purified | |
| RUO | |
| 26692 | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction