missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OR13C5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | OR13C5 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
OR13C5 Polyclonal specifically detects OR13C5 in Human samples. It is validated for Western Blot.Specifications
| OR13C5 | |
| Polyclonal | |
| Rabbit | |
| NP_001004482 | |
| 138799 | |
| Synthetic peptide directed towards the N terminal of human OR13C5. Peptide sequence ICYTTTSIPSTLVSFLSERKTISLSGCAVQMFLSLAMGTTECVLLGVMAF. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| olfactory receptor 13C5, olfactory receptor, family 13, subfamily C, member 5 | |
| OR13C5 | |
| IgG | |
| 36 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title