missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Optima™ 624 GC Capillary Column

Brand:  Macherey-Nagel™ 726787.25

Code : 50

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 10705962

  • £545.00 / Each
Estimated Shipment: 03-07-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

0.32 mm
GC Capillary Column
Optima™ 624
280°C to 300°C
25 m
G1, G2, G38
0.5 mm
Gas Chromatography (GC)
1.8 μm
1 Each
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers