missing translation for 'onlineSavingsMsg'
Learn More

Bollé Safety Optical Insert, Tracker Safety Goggles

Product Code. 12689210
Click to view available options
:
Clear
This item is not returnable. View return policy

Product Code. 12689210

Brand: Bollé Safety SOSTRACKER

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Ultra-comfortable, technical, and provides protection from all mechanical and chemical risks.

  • Optical insert.
  • Upper protection.
  • Removable foam reinforcement with indirect ventilation.
  • Lower protection.
  • Removable adjustable strap.
  • Side ventilation.
  • Platinum™ coating.
TRUSTED_SUSTAINABILITY

Specifications

Description Model SOSTRACKER
Color Clear
For Use With (Equipment) TRACKER Safety Goggles
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.