missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Opsin 1 (Medium Wave) Antibody - BSA Free, Novus Biologicals™
Chicken Polyclonal Antibody
£137.00 - £443.00
Specifications
| Antigen | Opsin 1 (Medium Wave) |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500:1:2000, ELISA 1:100-1:2000 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18119533
|
Novus Biologicals
NBP2-29858-0.025MG |
0.025 mg |
£137.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18179093
|
Novus Biologicals
NBP2-29858 |
0.1 mg |
£443.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Opsin 1 (Medium Wave) Polyclonal specifically detects Opsin 1 (Medium Wave) in Mouse, Rat samples. It is validated for Western Blot, ELISA.Specifications
| Opsin 1 (Medium Wave) | |
| Western Blot 1:500:1:2000, ELISA 1:100-1:2000 | |
| Polyclonal | |
| Chicken | |
| Vision | |
| PBS with 0.02% Sodium Azide | |
| GCP, GOP, Green cone photoreceptor pigment, Green-sensitive opsin, medium-wave-sensitive opsin 1, OPN1MW, opsin 1 (cone pigments), medium-wave-sensitive 2 | |
| OPN1MW | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| 1 mg/mL | |
| ELISA, Western Blot | |
| Unconjugated | |
| RUO | |
| Rat, Mouse | |
| P04000 | |
| 2652 | |
| Synthetic peptides from mouse green opsin and zebra finch red opsin. | |
| Affinity Purified | |
| Opsin, red and green. The antibody reacts with a protein of ∼40 kDa. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title