missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Opsin 1 (Medium Wave) Antibody - BSA Free, Novus Biologicals™
Chicken Polyclonal Antibody
Brand: Novus Biologicals NBP2-29858-0.025MG
This item is not returnable.
View return policy
Description
Opsin 1 (Medium Wave) Polyclonal specifically detects Opsin 1 (Medium Wave) in Mouse, Rat samples. It is validated for Western Blot, ELISA.
Specifications
| Opsin 1 (Medium Wave) | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| GCP, GOP, Green cone photoreceptor pigment, Green-sensitive opsin, medium-wave-sensitive opsin 1, OPN1MW, opsin 1 (cone pigments), medium-wave-sensitive 2 | |
| Chicken | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Opsin, red and green. The antibody reacts with a protein of ∼40 kDa. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| ELISA, Western Blot | |
| 1 mg/mL | |
| Western Blot 1:500:1:2000, ELISA 1:100-1:2000 | |
| P04000 | |
| OPN1MW | |
| Synthetic peptides from mouse green opsin and zebra finch red opsin. | |
| 0.025 mg | |
| Vision | |
| 2652 | |
| Rat, Mouse |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction