missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals ONO-RS-082, 98%

Product Code. 15458739
Change view
Click to view available options
Quantity:
20 mg
Unit Size:
20mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15458739 20 mg 20mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15458739 Supplier Thermo Scientific Chemicals Supplier No. J67444.F+
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 20 mg
Description Off-white to yellow
CAS 99754-06-0
Molecular Formula C21H22ClNO3
Formula Weight 371.86
Synonym 4-Chloro-2-(4-n-pentylcinnamoylamino)benzoic acid;
Sensitivity Light Sensitive
Melting Point 175°C to 177°C
Product Type ONO-RS-082, 98%
Form Solid
Assay 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.