missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oncostatin M/OSM Antibody (31E7), Novus Biologicals™
Human Monoclonal Antibody
£260.00 - £406.00
Specifications
| Antigen | Oncostatin M/OSM |
|---|---|
| Clone | 31E7 |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30026950
|
Novus Biologicals
NBP3-26445-50ul |
50 μL |
£260.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30037481
|
Novus Biologicals
NBP3-26445-100ul |
100 μL |
£406.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Oncostatin M/OSM Monoclonal antibody specifically detects Oncostatin M/OSM in Human samples. It is validated for ELISASpecifications
| Oncostatin M/OSM | |
| ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| MGC20461, oncostatin M, oncostatin-M | |
| Recombinant Human Oncostatin M/OSM protein [UniProt P13725] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 31E7 | |
| ELISA | |
| Unconjugated | |
| Human | |
| Biologically Active Proteins, Cell Cycle and Replication | |
| 50% Glycerol, 0.01M PBS, PH 7.4 | |
| 5008 | |
| IgG1 | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title