missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oncostatin M/OSM Antibody (31E7), Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP3-26445-100ul
This item is not returnable.
View return policy
Description
Oncostatin M/OSM Monoclonal antibody specifically detects Oncostatin M/OSM in Human samples. It is validated for ELISA
Specifications
| Oncostatin M/OSM | |
| Monoclonal | |
| Unconjugated | |
| 50% Glycerol, 0.01M PBS, PH 7.4 | |
| Human | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 31E7 | |
| ELISA | |
| MGC20461, oncostatin M, oncostatin-M | |
| Recombinant Human Oncostatin M/OSM protein [UniProt P13725] | |
| 100 μL | |
| Biologically Active Proteins, Cell Cycle and Replication | |
| 5008 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction