Learn More
Hamilton™ On-Column Injection Syringes with Removable Needles
Useful for the analysis of thermally unstable samples and avoids boiling point discrimination.
Brand:  Hamilton™ 87405
Code : 04
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 11520082
  • £133.00 / Each
Estimated Shipment: 11-07-2024
to see stock.
Description
Description
  • On-column injection employs the direct introduction of the liquid sample into the column
  • Uses point style 3 syringes
  • Point style 3 adapted to the typical septum of the on-column injector
  • Needles adjusted to the inner diameter of the column
Specifications
Specifications
Borosilicate Glass, PTFE
Gastight Syringe
1/Ea.
3
6.6 mm
1800 Series
10 μL
Removable