missing translation for 'onlineSavingsMsg'
Learn More

Diba Omnifit™ Replacement Filters for Glass Bottles

Solvent frits and filters for the bottom of the bottle remove even small particulates

Brand:  Diba

Code : Z3

Additional Details : Weight : 0.30000kg

 View more versions of this product

Product Code. 15204589

  • POA / Each
Estimated Shipment: 23-05-2024
to see stock.

This item requires a price update prior to being ordered.

Request a Price for this item here.


Description

Description

  • All-PTFE construction enables the filters to be used with harsh chemicals
  • The filter units have slots in the body to enable virtually all liquid to be removed from the bottle, minimizing waste

Specifications

Specifications

GL45 Bottle
10 μm
Each
0.31 cm
PTFE
To be used with the harsh chemicals
Replacement Filter
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers