Learn More
Diba Omnifit™ Replacement Filters for Glass Bottles
Solvent frits and filters for the bottom of the bottle remove even small particulates
POA
Specifications
Material PTFE
Pore Size 10 μm
Type Replacement Filter
View More Specs
Products 1
Product Code Brand For Use With (Equipment) No. per Case Price Quantity & Availability  
Product Code Brand For Use With (Equipment) No. per Case Price Quantity & Availability  
15204589
View Documents Promotion Details
Diba
009F-45
GL45 Bottle Each
POA
Each
View alternative products
Save Up to 
»
N/A
This item requires a price update prior to being ordered.
Request a Price for this item here.
Description
Description
  • All-PTFE construction enables the filters to be used with harsh chemicals
  • The filter units have slots in the body to enable virtually all liquid to be removed from the bottle, minimizing waste
Specifications
Specifications
PTFE
10 μm
Replacement Filter