Learn More
Diba Omnifit™ Luer Adapter
Adapters fit into 1/4 in.–28 UNF flat bottom ports in most Omnifit connectors and valves
POA
Product Code | Brand | Type | Material | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Material | Price | Quantity & Availability | ||||
15294479
|
Diba
002501 |
Male Luer Fitting | ETFE |
POA
Each |
N/A | ||||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
12786887
|
Diba
002502 |
Female Luer Fitting | ETFE |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Provide quick, push-on connection for softwall tubing or luer
- Made from chemically resistant ETFE
- Autoclavable
- OF001206 features a thread to male Luer lock adapter with a polypropylene lock ring
- OF002501 features male luer fittings to 1/4 in.–28 UNF thread
- OF002502 features femal luer fittings to 1/4 in.–28 UNF thread