missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Oleoyl-L-alpha-lysophosphatidic acid sodium salt

Product Code. p-7070108
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
This item is not returnable. View return policy

Product Code. 15408359

Brand: Thermo Scientific Chemicals J66836.#0

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 22556-62-3 | C21H40NaO7P, C21H40NaO7P | 458.51 g/mol

It is an endogenous agonist for LPA1 and LPA2 receptors. LPA does not induce angiogenesis, but has effects on endothelial cell physiology that are similar to those of sphingosine 1-phosphate. Induces cell migration of cancer and non-cancer cells. LPA has also been described to potentiate acetylchloine receptor currents by G-protein-mediated activation of Ca2+-dependent/independent protein kinase C.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22556-62-3
Molecular Formula C21H40NaO7P
Synonym 1-O-9Z-Octadecenoyl-sn-glyceryl-3-phosphoric acid sodium salt; Lysophosphatidic Acid

Specifications

Quantity 5 mg
Description White to off-white
Content And Storage -30°C to -10°C
Formula Weight 458.5
Sensitivity Air and Light Sensitive, Hygroscopic
Solubility Soluble in water up to 5mg/ml with sonication
Product Type Oleoyl-L-alpha-lysophosphatidic acid sodium salt
Form Waxy solid or powder
InChI Key XGRLSUFHELJJAB-UHFFFAOYNA-M
SMILES [Na+].CCCCCCCCC=CCCCCCCCC(=O)OCC(O)COP(O)([O-])=O
IUPAC Name sodium 3-(hydrogen phosphonatooxy)-2-hydroxypropyl octadec-9-enoate
Molecular Weight (g/mol) 458.51
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.