missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius™ OIML Class F2 Proof Line Knob Weight with DAkkS Certificate
Polished non-magnetizable calibration weights. Sartorius™ OIML Class F2 Proof Line Knob Weight with DAkkS Certificate is constructed of special steel and supplied in a plastic case.
£104.00 - £717.00
Specifications
Certifications/Compliance | DAkkS calibration certificate |
---|---|
Classification | OIML Class F2 |
Density | 7,950kg/m3 |
Product Code | Brand | Mass | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Mass | Price | Quantity & Availability | |||||
11373384
|
Sartorius™
YCW314-AC-02 |
1 g |
£105.00
Each |
In Stock
Log in to see stock. |
|||||
11313394
|
Sartorius™
YCW324-AC-02 |
2 g |
£104.00
Each |
Estimated Shipment: 02-07-2024
Log in to see stock. |
|||||
11353394
|
Sartorius™
YCW354-AC-02 |
5 g |
£107.00
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
11393394
|
Sartorius™
YCW414-AC-02 |
10 g |
£110.00
Each |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
11333404
|
Sartorius™
YCW424-AC-02 |
20 g |
£110.00
Each |
Estimated Shipment: 02-07-2024
Log in to see stock. |
|||||
11373404
|
Sartorius™
YCW454-AC-02 |
50 g |
£116.00
Each |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
11313414
|
Sartorius™
YCW514-AC-02 |
100 g |
£123.00
Each |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||||
11353414
|
Sartorius™
YCW524-AC-02 |
200 g |
£136.00
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||||
11393414
|
Sartorius™
YCW554-AC-02 |
500 g |
£190.00
Each |
Estimated Shipment: 02-07-2024
Log in to see stock. |
|||||
11333424
|
Sartorius™
YCW614-AC-02 |
1 kg |
£203.00
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||||
11373424
|
Sartorius™
YCW624-AC-02 |
2 kg |
£259.00
Each |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
11313434
|
Sartorius™
YCW654-AC-02 |
5 kg |
£388.00
Each |
Estimated Shipment: 27-06-2024
Log in to see stock. |
|||||
11397494
|
Sartorius™
YCW714-AC-02 |
10 kg |
£717.00
Each |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
Description
- Non-magnetizable
- Supplied in a high-quality plastic case (unless otherwise specified) suitable for use in cleanrooms
- Special Stainless steel construction with a density of 7,950 kg/m3
- Complies with OIML R111-1 International Recommentation
- DAkkS Certified
Specifications
DAkkS calibration certificate | |
OIML Class F2 | |
7,950kg/m3 |