missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Oil Test Paper

Product Code. 10132251 Shop All Macherey-Nagel Products
Change view
Click to view available options
Color Indicator:
Pale Blue to Dark Blue
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Indicator unitSize
10132251 Pale Blue to Dark Blue Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10132251 Supplier Macherey-Nagel™ Supplier No. 90760

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Allows the rapid, reliable and qualitative detection of oil contaminations of water and soil. Macherey Nagel™ Oil Test Papers' sensitivity largely depends on the nature of the respective hydrocarbon.

  • For soil detection: press paper firmly against soil to be tested and rinse with clear water
  • For water detection: move paper back and forth in the sample a few times
  • In case of volatile hydrocarbons, color reaction of test paper has to be immediately evaluated

Specifications

Color Indicator Pale Blue to Dark Blue
Format Strip
For Use With (Application) Determination of oil contaminations
Quantity 100 Pack
Type Qualitative Test Paper
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.