missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Odf1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00 - £366.00
Specifications
| Antigen | Odf1 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18659650
|
Novus Biologicals
NBP2-93601-0.02ml |
0.02 mL |
£151.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18606580
|
Novus Biologicals
NBP2-93601-0.1ml |
0.1 mL |
£366.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Odf1 Polyclonal antibody specifically detects Odf1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| Odf1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| cancer/testis antigen 133, CT133, HSPB10, MGC129928, MGC129929, ODF, ODF2, ODF27, ODFP, ODFPG, ODFPGA, ODFPGB, outer dense fiber of sperm tails 1, outer dense fiber of sperm tails, 27-kD, outer dense fiber protein 1, outer dense fibre of sperm tails 1, RT7, SODF | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 80-180 of human ODF1 (NP_077721.2). LYCLRPSLRSLERKAIRAIEDEKRELAKLRRTTNRILASSCCSSNILGSVNVCGFEPDQVKVRVKDGKVCVSAERENRYDCLGSKKYSYMNICKEFSLPPC | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 4956 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title