missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Odf1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93601-0.02ml
This item is not returnable.
View return policy
Description
Odf1 Polyclonal antibody specifically detects Odf1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| Odf1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| cancer/testis antigen 133, CT133, HSPB10, MGC129928, MGC129929, ODF, ODF2, ODF27, ODFP, ODFPG, ODFPGA, ODFPGB, outer dense fiber of sperm tails 1, outer dense fiber of sperm tails, 27-kD, outer dense fiber protein 1, outer dense fibre of sperm tails 1, RT7, SODF | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 80-180 of human ODF1 (NP_077721.2). LYCLRPSLRSLERKAIRAIEDEKRELAKLRRTTNRILASSCCSSNILGSVNVCGFEPDQVKVRVKDGKVCVSAERENRYDCLGSKKYSYMNICKEFSLPPC | |
| 0.02 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 4956 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction