missing translation for 'onlineSavingsMsg'
Learn More

Octylmagnesium bromide, 2M solution in diethyl ether,AcroSeal™

Product Code. 10295204 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
800mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10295204 800 mL AcroSeal™ Glass Bottle 800mL
10235164 100 mL AcroSeal™ Glass Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10295204 Supplier Thermo Scientific Acros Supplier No. 427818000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 17049-49-9 | C8H17BrMg | 217.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17049-49-9 , 60-29-7
Molecular Formula C8H17BrMg
Molecular Weight (g/mol) 217.43
MDL Number MFCD00191566
InChI Key ZDUZWGUHIRQPGP-UHFFFAOYSA-M
Synonym bromooctyl magnesium, n-octylmagnesium bromide, magnesium, bromooctyl, octylmagnesium bromide, n-octyl magnesium bromide, n-octylmagnesiumbromide, octyl magnesium bromide, c8h17mgbr, 1-octylmagnesium bromide, octylmagnesium bromide solution, grignard reagent
PubChem CID 86923
IUPAC Name bromo(octyl)magnesium
SMILES CCCCCCCC[Mg]Br

Specifications

Boiling Point 35.0°C
Color Yellow to Brown
CAS Min % 42.99, 50.22
CAS Max % 49.78, 57.01
Physical Form Liquid
Quantity 800 mL
Linear Formula CH3(CH2)7MgBr
Flash Point −35°C
Formula Weight 217.43
Concentration or Composition (by Analyte or Components) 1.90 to 2.20M
Packaging AcroSeal™ Glass Bottle
Specific Gravity 0.961
Density 0.9610g/mL
Chemical Name or Material Octylmagnesium bromide, 2M solution in diethyl ether
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Octylmagnesium bromide, 2M solution in diethyl ether
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H224-Extremely flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P243-Take precautionary measures against static discharge.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.
  • P403+P235-Store in a well-ventilated place. Keep cool.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.