CAS RN 17049-49-9
CAS RN 17049-49-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
217.43
Molecular Formula:
C8H17BrMg
InChi Key:
IOOQQIVFCFWSIU-UHFFFAOYSA-M
SMILES:
[Mg++].[Br-].CCCCCCC[CH2-]
1
–
1
of
1
results
1
Octylmagnesium bromide, 2M solution in diethyl ether,AcroSeal™
CAS: 17049-49-9 Molecular Formula: C8H17BrMg Molecular Weight (g/mol): 217.43 MDL Number: MFCD00191566 InChI Key: ZDUZWGUHIRQPGP-UHFFFAOYSA-M Synonym: bromooctyl magnesium,n-octylmagnesium bromide,magnesium, bromooctyl,octylmagnesium bromide,n-octyl magnesium bromide,n-octylmagnesiumbromide,octyl magnesium bromide,c8h17mgbr,1-octylmagnesium bromide,octylmagnesium bromide solution,grignard reagent PubChem CID: 86923 IUPAC Name: bromo(octyl)magnesium SMILES: CCCCCCCC[Mg]Br