missing translation for 'onlineSavingsMsg'
Learn More

Octinoxate, TRC

Product Code. 30135041
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135041 10 g 10g
30132582 100 g 100g
30009861 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135041 Supplier Toronto Research Chemicals Supplier No. TRCO25750010G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5466-77-3
Molecular Formula C18 H26 O3
Molecular Weight (g/mol) 290.4
Synonym 2-Ethylhexyl 4-methoxycinnamate, 2-Ethylhexyl methoxycinnamate, 2-Ethylhexyl p-methoxycinnamate, 3-(4-Methoxyphenyl)-2-propenoic acid 2-ethylhexyl ester, 4-Methoxycinnamic acid 2-ethylhexyl ester, Escalol 557, Escalol 557NB, Escalol 557T, Ethylhexyl methoxycinnamate, Ethylhexyl p-methoxycinnamate, Eusolex 2292, Eusolex UV-Pearls OMC, Jeescreen OMC, NSC 26466, Neo Heliopan AV, Nomcort TAB, Nomcort TAB-R, Octinoxate, Octyl 4-methoxycinnamate, Octyl methoxycinnamate, Octyl p-methoxycinnamate, Parsol MCX, Parsol MCX-SA, Parsol MOX, Solarom OMC, Sun Caps 664, Sunscreen AV, Tinosorb OMC, Uvinul 3088, Uvinul MC 80, Uvinul MC 80N, Uvinul MC 90, p-Methoxycinnamic acid 2-ethylhexyl ester
IUPAC Name 2-ethylhexyl 3-(4-methoxyphenyl)prop-2-enoate
SMILES CCCCC(CC)COC(=O)C=Cc1ccc(OC)cc1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C18H26O3/c1-4-6-7-15(5-2)14-21-18(19)13-10-16-8-11-17(20-3)12-9-16/h8-13,15H,4-7,14H2,1-3H3
Formula Weight 290.1882
Percent Purity >95%
Chemical Name or Material Octinoxate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.