missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OCT1 Rabbit anti-Human, Porcine, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10387-1-AP-150UL
This item is not returnable.
View return policy
Description
The OCT1 transcription factor was among the first identified members of the POU transcription factor family. Members of this family contain the POU domain, a 160-amino acid region necessary for DNA binding to the octameric sequence ATGCAAAT.Specifications
| OCT1 | |
| Polyclonal | |
| Unconjugated | |
| POU2F1 | |
| NF A1, Oct 1, OCT1, Octamer binding protein 1, OTF 1, OTF1, POU class 2 homeobox 1, POU2F1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 397501, 5451 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P14859, Q29076 | |
| POU2F1 | |
| OCT1 Fusion Protein Ag0586 | |
| 150 μL | |
| Primary | |
| Human, Pig | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction