missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OCT1 Rabbit anti-Human, Porcine, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | OCT1 |
|---|---|
| Concentration | 0.23 mg/mL |
| Applications | Immunoprecipitation, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16846743
|
Proteintech
10387-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16836743
|
Proteintech
10387-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The OCT1 transcription factor was among the first identified members of the POU transcription factor family. Members of this family contain the POU domain, a 160-amino acid region necessary for DNA binding to the octameric sequence ATGCAAAT.Specifications
| OCT1 | |
| Immunoprecipitation, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| Pig, Human | |
| P14859, Q29076 | |
| 397501, 5451 | |
| OCT1 Fusion Protein Ag0586 | |
| Primary | |
| -20°C | |
| POU2F1 |
| 0.23 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| NF A1, Oct 1, OCT1, Octamer binding protein 1, OTF 1, OTF1, POU class 2 homeobox 1, POU2F1 | |
| POU2F1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title