missing translation for 'onlineSavingsMsg'
Learn More

Occludin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen Occludin
Concentration 0.27 mg/mL
Applications Western Blot, Immunocytochemistry, Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16801614
View Documents
Proteintech
13409-1-AP-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16891604
View Documents
Proteintech
13409-1-AP-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This gene encodes an integral membrane protein that is required for cytokine-induced regulation of the tight junction paracellular permeability barrier. Mutations in this gene are thought to be a cause of band-like calcification with simplified gyration and polymicrogyria (BLC-PMG), an autosomal recessive neurologic disorder that is also known as pseudo-TORCH syndrome. Alternative splicing results in multiple transcript variants. A related pseudogene is present 1.5 Mb downstream on the q arm of chromosome 5.
Specifications

Specifications

Occludin
Western Blot, Immunocytochemistry, Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Rat, Mouse, Human
Q16625, Q61146, Q6P6T5
100506658, 18260, 83497
Occludin Fusion Protein Ag4057
Primary
-20°C
OCLN
0.27 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
occludin, OCLN
Ocln
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.